Learn More
Medchemexpress LLC Tizaterkib | 2097416-76-5 | MFCD31631580 | 99.1% | 494.50 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111483
Encompass_Preferred
Tizaterkib (AZD0364) is a potent and selective ERK2 inhibitor, with an IC50 of 0.6 nM. It is intended for research use only and not for patients.
  • Potent and selective ERK2 inhibitor
  • Inhibits the growth of cancer cell lines with KRAS mutations as monotherapy
  • Enhances effect synergistically when combined with Selumetinib
  • Reduces tumor growth in A549 xenograft models in combination with MEK inhibitor Selumetinib
Catalog No. 50-003-46812