Learn More
Cayman Chemical TIzoxnIde-d4 1mg
SDP
Supplier:  Cayman Chemical 285331 MG
Encompass_Preferred
An internal standard for the quantification of tizoxanide by GC- or LC-MS
Catalog No. 50-294-0551