Learn More
Medchemexpress LLC TK-112690 | 22423-26-3 | 240.21 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00849150G
Encompass_Preferred
TK-112690 is a UPP1 inhibitor that can be used for the study of cancer and pulmonary fibrosis. It inhibits murine small intestinal uridine phosphorylase (UPase) with an IC50 of 12.5 μM and human small intestinal UPase with an IC50 of 20.0 μM in vitro. This product also increases plasma uridine concentration in mice.
  • Inhibits murine small intestinal uridine phosphorylase (UPase) with an IC50 of 12.5 μM
  • Inhibits human small intestinal UPase with an IC50 of 20.0 μM in vitro
  • Increases plasma uridine concentration in mice
  • Used for the study of cancer and pulmonary fibrosis
Catalog No. 50-004-47288