Learn More
Medchemexpress LLC Tk-129 | 3031476-73-7 | C15H23N5O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15148310MM/1ML
Encompass_Preferred
TK-129 is an orally active, low-toxicity, and potent KDM5B inhibitor with a high affinity (IC50=44 nM). It demonstrates cardioprotective effects by inhibiting KDM5B and interfering with the KDM5B-associated Wnt pathway. In vitro studies, TK-129 reduces ang II-induced activation of cardiac fibroblasts. In vivo, it has been shown to reduce isoprenaline-induced myocardial remodelling and fibrosis. This compound is suitable for use in studies related to cardiovascular disease.
  • Orally active
  • Low-toxicity
  • Potent KDM5B inhibitor
  • High affinity (IC50=44 nM)
  • Cardioprotective effects
  • Inhibits KDM5B and blocks the KDM5B-associated Wnt pathway
  • Reduces ang ii-induced activation of cardiac fibroblasts in vitro
  • Reduces isoprenaline-induced myocardial remodelling and fibrosis in vivo
  • Suitable for cardiovascular disease studies
Catalog No. 50-003-72190