Learn More
ATCC TK6; Lymphoblast; Human (Homo sapiens)
Supplier:  ATCC CRL8015
Encompass
TK6 is a lymphoblast cell line that was isolated from the spleen of a 5-year-old, male with hereditary spherocytosis. The cells are heterozygous at the thymidine kinase (TK) locus and can be used to quantitatively detect forward mutations at 3 loci (resistance to trifluorothymidine (tk locus)).
Catalog No. 50-238-3209
 Restricted Item