Learn More
Hach Company TKN Indicator Solution, 50 mL SCDB
Supplier:  Hach Company 2251926
Encompass
Indicator solution for pH adjustment in Total Kjeldahl Nitrogen determination by the Nessler Method. 50 mL Self-Contained Dropping Bottle
Catalog No. 50-199-5635