Learn More
Medchemexpress LLC TL-895 | 1415823-49-2 | 99.9% | 447.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139481100MG
Encompass_Preferred
TL-895 is a potent, orally active, ATP-competitive, and highly selective irreversible BTK inhibitor. It is active against recombinant BTK (average IC50: 1.5 nM) and inhibits only a few other kinases like BLK, BMX, and TXK. This product inhibits BTK auto-phosphorylation at the Y223 phosphorylation site (IC50: 1-10 nM) and effectively reduces the production of inflammatory factors (IL-8, IL-1β, MCP-1, TNF-α) by monocytes or macrophages. It also reduces the chemotactic migration of MF cells towards SDF-1. TL-895 is utilized in studies of chronic lymphocytic leukemia (CLL), myelofibrosis (MF), and B-cell malignancies.
  • Potent, orally active, ATP-competitive, and highly selective irreversible BTK inhibitor
  • Active against recombinant BTK
  • Inhibits BTK auto-phosphorylation
  • Reduces production of inflammatory factors
  • Reduces chemotactic migration of MF cells
  • Used for studies of CLL, MF, and B-cell malignancies
Catalog No. 50-003-51361