Learn More
Medchemexpress LLC Tl13-112 | 2229037-19-6 | 99.2% | 1002.57 g·mol⁻1 | C49H60ClN9O10S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12391925MG
Encompass_Preferred
TL13-112 is a bifunctional PROTAC small molecule that promotes proteasomal degradation of anaplastic lymphoma kinase (ALK) by recruiting an E3 ligase. It is intended for use in biochemical and cell-based studies of targeted protein degradation and kinase biology, with reported potent ALK activity and cellular degradation profiles.
  • Induces ALK degradation in cell-based assays.
  • Potent ALK activity with low-nanomolar inhibitory concentration.
  • Designed for mechanistic studies of targeted protein degradation.
  • Soluble in common organic solvents for assay preparation.
  • High reported purity suitable for research applications.
Catalog No. 50-002-36041