Learn More
Medchemexpress LLC TL13-112 | 2229037-19-6 | MFCD12922943 | 98.0% | 1002.57 g/mol | C49H60ClN9O10S | 100MG

Supplier: Medchemexpress LLC HY123919100MG
TL13-112 is a potent, selective anaplastic lymphoma kinase (ALK) PROTAC degrader supplied for research use. It inhibits ALK activity and induces ALK degradation in cellular assays; analytical data, certificates, and stability information are provided by the supplier.
- Potent ALK degrader with reported ALK IC50 of 0.14 nM.
- Molecular weight 1002.57 g/mol and formula C49H60ClN9O10S.
- Purity reported as 98.0% with supporting analytical data.
- Soluble in DMSO at approximately 100 mg/mL.
- Provided as a solid with recommended storage conditions for powder and in solvent.
- Intended for biochemical and cell-based research applications only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.