Learn More
Medchemexpress LLC TL13-68 | 98.9% | 851.46 g/mol | C41H55ClN10O6S | 5 MG

Supplier: Medchemexpress LLC HY1368495MG
TL13-68 is a biotin-tagged derivative of SM1-71 provided for research use. It functions as an affinity probe to study the mechanism and molecular interactions of SM1-71 in biochemical and cellular assays. The compound is supplied as a purified small-molecule powder and should be handled under standard laboratory safety procedures (research use only).
- Biotin tag enables affinity capture and streptavidin-based assays.
- High purity (98.9%) suitable for biochemical studies.
- Molecular weight 851.46 g/mol; molecular formula C41H55ClN10O6S.
- Supplied as a solid powder for flexible formulation.
- Store at 4°C under inert atmosphere; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.