Learn More
Cayman Chemical TLFTBP lIke factr Monclo 100uL
SDP
Supplier:  Cayman Chemical 33113100 UL
Encompass_Preferred
A sesquiterpene lactone with diverse biological activities inhibits mycelial growth in the phytopathogenic fungi B. cinerea F. coeruleum C. lunata F. graminearum and A. brassicae by 33.93 67.14 27.04 46.22 and 32.48 respectively at 100 g/ml reduces infection of HEK293T cells by an HIV-based virus system pseudotyped with Ebola virus glycoprotein (EC50 8 M) inhibits nitric oxide production in LPS-stimulated BV-2 cells (IC50 20.3 M) enhances gemcitabine-induced cell death in gemcitabine-resistant PANC-1 and AsPC-1 human pancreatic cancer cells at 5 10 and 20 M
Catalog No. 50-294-8467