Learn More
Enzo Life Sciences TLR2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905710100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human TLR2. UniProt ID: O60603. Species reactivity: Human, Mouse. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (2µg/ml) Western Blot (0.5-2.0µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ion exchange purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7003