Learn More
Invivogen TLR4 LIGAND E.COLI LPS
SDP
Supplier:  Invivogen TLRLEBLPS
Encompass
Purified Lipopolysaccharide, TLR4 Ligands, E. coli LPS, Working Concentration: 10 ng to 10 microgram/mL, Quantity: 5 mg
Catalog No. NC9039780