Learn More
Selleck Chemical LLC TLR7/8 agonist 1 2HCl
SDP
Supplier:  Selleck Chemical LLC E06395MG
Encompass_Preferred
TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline TLR7/8 agonist 1 retains TLR7-agonistic activity with an EC50 of 20 nM
Catalog No. 50-313-7021