Learn More
Medchemexpress LLC TLR7 agonist 3 | 1229024-78-5 | 99.9% | 312.41 | C18H24N4O | 10 MG

Supplier: Medchemexpress LLC HY11760210MG
TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 used for preclinical in-vitro and in-vivo research. It is supplied as a white to off-white solid with high reported purity, defined solubility, and recommended storage to support formulation and dosing.
- Potent TLR7 agonist for immune activation studies.
- White to off-white solid appearance.
- High purity (99.9%).
- Molecular formula C18H24N4O; molecular weight 312.41 g/mol.
- Soluble in DMSO and ethanol; compatible with co-solvent in vivo formulations.
- Recommended storage: powder at -20°C for long term, 4°C for shorter storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.