Learn More
Aviva Systems Biology Corp TLX3 Antibody - N-terminal region (ARP31925_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31925_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLX3. Alternate names: HOX11L2, MGC29804, RNXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54374