Learn More
Medchemexpress LLC TLyP-1 peptide | 1354801-55-0 | 98.1% | 833.96 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1078710MG
Encompass_Preferred
tLyP-1 peptide is an NRP-1 targeting peptide with an IC50 of 4 μM. Its amino acid sequence is CGNKRTR, and it specifically binds to NRP-1 to target tumor cells. This product is for research use only.
  • NRP-1 targeting peptide
  • IC50 of 4 μM
  • Amino acid sequence: CGNKRTR
  • Specifically binds to NRP-1
  • Targets tumor cells
  • For research use only
Catalog No. 50-004-23071