Learn More
Medchemexpress LLC TM4SF2/TSPAN7 Cynom 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP77495500UG
Encompass_Preferred
TM4SF2/TSPAN7 protein is an important member of the tetraspanin family and plays an important role in signal transduction cell adhesion and membrane organization Its research enhances understanding of cell membrane dynamics and provides potential applications in cell biology and cancer research TM4SF2/TSPAN7 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived TM4SF2/TSPAN7 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-86936