Learn More
Medchemexpress LLC TMA chloride, for molecular biology | 75-57-0 | 99.0% | 109.60 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW012954B10G
Encompass_Preferred
TMA chloride is a chemical reagent used in molecular biology. It can increase the specificity of hybridization and raise the DNA denaturation temperature in PCR, making it suitable for various research applications.
  • Increases specificity of hybridization
  • Raises DNA denaturation temperature in PCR
  • Used as a biochemical assay reagent
  • Suitable for laboratory chemical manufacturing
  • For research use only
Catalog No. 50-004-54084