Learn More
Abcam TMB ELISA Substrate (Fast Kinetic Rate), 1000ML
Supplier:  Abcam AB1715241000ML
Encompass_Preferred
Abcam's TMB ELISA Substrate (Fast Kinetic Rate) detects horseradish peroxidase (HRP) activity and contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2490