Learn More
Abcam TMB ELISA Substrate (Slowest Kinetic Rate), 100ML
Supplier:  Abcam AB171527100ML
Encompass_Preferred
Abcam's TMB ELISA Substrate (Slowest Kinetic Rate) contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1980316