Learn More
Sigma Aldrich Fine Chemicals Biosciences TMB Enhanced One Component1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T94551L
Encompass
TMB is a substrate for horseradish peroxidase. The substrate system will react with sites on the membrane containing peroxidase producing an insoluble permanent dark blue reaction product. To stop the reaction rinse the membrane with reagent quality water. This substrate is not recommended for microwell or immunohistochemical applications.
Catalog No. 50-291-4133