Learn More
Surmodics Inc TMB Enhanced HRP Membrane Substrate 100ml
SDP
Supplier:  Surmodics Inc ESPM010001
Encompass_Preferred
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. A highly intense staining pattern increases the signal-to-noise ratios for improved sensitivity in western blot, dot blots and line assays.
Catalog No. NC1306054