Learn More
Seracare Life Sciences Inc TMB MEMBRANE ENHANCER
Supplier:  Seracare Life Sciences Inc 54200026
Encompass_Preferred
Component of the TMB Membrane Peroxidase Substrate System which allows the substrate to precipitate. TMB Membrane Peroxidase Substrate develops a dark blue stain at the reaction site in the presence of peroxidase-labeled conjugates. The TMB Membrane Enhancer is recommended for use with KPL's 2-compenent TMB Microwell Substrate (Cat. No. 50-76-00) to produce a precipitating substrate.
Catalog No. 50-674-24