Learn More
Cayman Chemical TMBhydrochlorIde hydrate 250mg
SDP
Supplier:  Cayman Chemical 70450250 MG
Encompass_Preferred
A colorimetric peroxidase substrate; enzymatic oxidation produces a radical cation followed by a diimine product that can be quantified by colorimetric detection at 652 and 450 nm, respectively, as a measure of peroxidase activity; has commonly been used as a substrate for HRP in ELISA and immunohistochemical applications
Catalog No. 50-289-4189