Learn More
Aviva Systems Biology Corp TMED10 Antibody - middle region (ARP58728_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMED10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMED10. Alternate names: P24(DELTA), S31I125, S31III125, TMP21, Tmp-21-I, p23ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62405