Learn More
Medchemexpress LLC TMEM156 Human HEK2 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77244100UG
Encompass_Preferred
TMEM156 Protein is a member of transmembrane proteins (TMEM) family The expression levels of TMEM156 correlates with patient survival TMEM156 is upregulated in tumor tissue Many genes positively correlated with TMEM156 are involved in multiple immunological processes Furthermore genes negatively correlated with TMEM156 are linked with RHO GTPase effectors which results in a poorer response to receptor activation thus reducing cancer cell proliferation survival and migration TMEM156 Protein Human (HEK293 His) is the recombinant human-derived TMEM156 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86540