Learn More
Aviva Systems Biology Corp TMEM16C Antibody - middle region (ARP49996_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49996_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM16C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM16C. Alternate names: C11orf25, GENX-3947, TMEM16CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60234