Learn More
Medchemexpress LLC TMEM219/IGFBP-3R Hu 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7796220UG
Encompass_Preferred
The TMEM219/IGFBP-3R protein was identified as an IGFBP3-specific cell death receptor and a potential mediator of caspase-8-dependent apoptosis upon ligand binding This protein actively binds to IGFBP3 forming a key molecular interaction that may initiate the apoptosis signaling pathway TMEM219/IGFBP-3R Protein Human (HEK293 Fc) is the recombinant human-derived TMEM219/IGFBP-3R protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-95215