Learn More
Medchemexpress LLC TMEM219/IGFBP-3R Mo 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7769750UG
Encompass_Preferred
The IGFBP-3R protein was identified as an IGFBP3-specific cell death receptor and becomes a potential mediator of caspase-8-dependent apoptosis upon ligand binding This receptor actively interacts with IGFBP3 forming a key molecular connection that may initiate the apoptotic signaling cascade TMEM219/IGFBP-3R Protein Mouse (HEK293 Fc) is the recombinant mouse-derived IGFBP-3R protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87328