Learn More
Strem, An Ascensus Company CAS# 1445-79-0. 25g. Trimethylgallium, 99+%, 31-2000, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014841
SDP
Supplier:  Strem, An Ascensus Company 98406825G
Encompass
CAS# 1445-79-0. 25g. Trimethylgallium, 99+%, 31-2000, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014841. Molecular Weight: 114.83. Molecular Formula: (CH3)3Ga. Color/form: colorless liq. Strem# 98-4068. http://www.strem.com/catalog/v/98-4068/
Catalog No. 50-204-5453