Learn More
Enzo Life Sciences TMP-153 (100mg). CAS: 128831-46-9
SDP
Supplier:  Enzo Life Sciences BMLEI3170100
Encompass_Preferred
TMP-153 is a potent and selective inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT), IC50=5-10 nM. It inhibits cholesterol absorption and lowers plasma cholesterol in rats and hamsters. ACAT inhibitors improve plasma lipid profile and ameliorate proteinuria in nephrotic animals. Purity: ≥98%. Solubility: Soluble in DMSO (>25mg/ml) or ethanol (10mg/ml with warming). Long Term Storage: Ambient.
Catalog No. 50-201-0706