Learn More
Selleck Chemical LLC TMP195
SDP
Supplier:  Selleck Chemical LLC S850210MM/1ML
Encompass_Preferred
TMP195 (TFMO 2) is a selective first-in-class class IIa HDAC inhibitor with Ki of 59 60 26 and 15nM for HDAC4 HDAC5 HDAC7 and HDAC9 respectively
Catalog No. 50-000-01864