Learn More
Medchemexpress LLC Tmp780 | 1422053-03-9 | MFCD32878247 | 99.5% | 494.58 | C31H30N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY102075B50MG
Encompass_Preferred
TMP780 is a small-molecule inverse agonist of RORγt with a reported IC50 of approximately 13 nM. It is used in inflammation and immunology research to probe RORγt-mediated pathways and to support preclinical studies of cutaneous inflammatory disorders.
  • Inverse agonist of RORγt with reported IC50 ≈ 13 nM.
  • Suitable for in vitro and preclinical inflammation and immunology research.
  • High purity material (~99.5%) for biochemical and cellular assays.
  • Supplied as a 50 mg research pack for laboratory use.
Catalog No. 50-002-45572