Learn More
Medchemexpress LLC Tmprss6-in-1 TFA | 00-00-0 | >98.0% | 790.88 g·mol⁻¹ | C35H41F3N8O6S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY130136A5MG
Encompass_Preferred
TMPRSS6-IN-1 TFA is a research-grade small-molecule inhibitor of TMPRSS6 (matriptase-2), supplied as the trifluoroacetic acid (TFA) salt. It is intended for in vitro biochemical and cell-based studies of serine/threonine protease activity and pathways related to iron homeostasis. Supplied as a solid in small milligram quantities; for research use only.
  • Potent inhibitor of TMPRSS6 (matriptase-2).
  • Supplied as the trifluoroacetic acid (TFA) salt for formulation convenience.
  • High solubility in DMSO (125 mg/mL) enables concentrated stock solutions.
  • Available in small milligram quantities suitable for screening and assay development.
  • Store sealed and away from moisture; in solution: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-62866