Learn More
Apexbio Technology LLC TMPyP4 tosylate 36951-72-1 50mg
SDP
Supplier:  Apexbio Technology LLC B762750
Encompass_Preferred
TMPyP4 tosylate (CAS 36951-72-1) is a small-molecule inhibitor targeting nucleic acid G-quadruplex structures It is designed to stabilize G-quadruplexes thereby inhibiting telomerase enzyme activity and disrupting IGF-1 mediated signaling pathways to regulate cellular proliferation TMPyP4 tosylate exerts its biological activity primarily through selective binding to G-quadruplex nucleic acid structures In cell-based studies TMPyP4 demonstrates anti-telomerase activity with reported IC50 values typically ranging from 10 50 M depending on the cell line and experimental conditions Based on these pharmacological properties TMPyP4 tosylate holds research potential in cancer biology and antiviral therapeutic studies including applications against SARS-CoV-2
Catalog No. 50-000-91679