Learn More
Cayman Chemical TMS Fatty Acids and Derivative
SDP
Supplier:  Cayman Chemical 1003810 MG
Encompass_Preferred
CYP1B1 is mainly an extrahepatic enzyme which oxidatively metabolizes both endogenous (steroids; eicosanoids) and exogenous xenobiotics such as polyaromatic hydrocarbons. TMS is a potent and selective inhibitor of CYP1B1, with an IC50 of 6 nM.{10511} It is 50-fold selective for the inhibition of CYP1B1 versus CYP1A1, making it a useful tool to differentiate between various CYP450 families.{10511} In cultured human colon cancer cells, TMS induces apoptosis and inhibits cell growth with an IC50 of 0.8 UG/ml.{10510}
Catalog No. 50-288-9799