Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions TMS Solution, clear liquid, 1 L, CAS: 142-82-5, 110-86-1, density 0.791, boiling point 98°C, shelf life 6 months
SDP

Supplier:  Aqua Solutions SPE618A

Encompass_Preferred

The AQUA SOLUTIONS TMS Solution (SPE618A) is a precisely characterized clear liquid, supplied in a convenient 1 L volume, suitable for diverse laboratory applications. This solution has specific CAS numbers (142-82-5, 110-86-1) and features a density of 0.79131 g/mL with a boiling point of 98°C. Designed for scientific use, it provides a reliable shelf life of 6 months. One important characteristic for handling is its limited miscibility with water, making it an essential choice for specialized experiments.

  • Clear liquid form
  • Volume of 1 L
  • CAS numbers 142-82-5, 110-86-1
  • Density of 0.79131 g/mL
  • Boiling point of 98°C
  • Not miscible or difficult to mix with water
  • Shelf life of 6 months

Catalog No. NC2146353


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.