Learn More
Selleck Chemical LLC TMZ(Temozolomide) S1237-1g
SDP
Supplier:  Selleck Chemical LLC S12371G
Encompass_Preferred
TMZ(Temozolomide) is a monofunctional SN-1 alkylating agent that can modify nitrogen atoms in the DNA ring and the extracyclic oxygen group chemically converted to MTIC and degrades to methyldiazonium cation which transfers methyl groups to DNA at physiologic pH A DNA damage inducer in L-1210 and L-1210/BCNU cells Temozolomide induces apoptosis and exhibits antitumor activity
Catalog No. 50-311-1207