Learn More
Abcam TNB, Yellow chromogenic standard for DTNB thiol quantification, 50MG
Supplier:  Abcam AB14563250MG
Encompass_Preferred
MW 199.19 Da, Purity =95%. TNB, Yellow chromogenic standard for DTNB thiol quantification. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4978