Learn More
Abcam TNBT, Redo x indicator, 1G
Supplier:  Abcam AB1462631G
Encompass_Preferred
MW 907.6 g/mol. Redox indicator for enzymes. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4782