Learn More
Medchemexpress LLC TNF-α (31-45), human | 144796-71-4 | 99.99% | 1667.87 | 5 MG

Supplier: Medchemexpress LLC HYP18605MG
TNF-α (31-45), human is a potent NF-kB pathway activator and a proinflammatory cytokine that induces necrosis or apoptosis. It stimulates the NF-κB pathway via TNFR2, promoting cancer growth, invasion, and metastasis.
- Potent NF-κB pathway activator
- Induces necrosis or apoptosis
- Stimulates NF-κB pathway via TNFR2
- Promotes cancer growth, invasion, and metastasis
- High purity (99.99% by HPLC)
- Molecular weight of 1667.87
- White to off-white solid appearance
- Sequence: Arg-Arg-Ala-Asn-Ala-Leu-Leu-Ala-Asn-Gly-Val-Glu-Leu-Arg-Asp
- Soluble in water at 50 mg/mL (requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.