Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Mo 100ug
SDP
Supplier:  Medchemexpress LLC HYP70516100UG
Encompass_Preferred
Recombinant mouse TNF RII (TNFRSF1B) extracellular domain expressed in HEK293 cells with a C-terminal 6xHis tag, provided as purified protein for research use. It binds tumor necrosis factor-α and is used to study TNF signaling, receptor crosstalk, TRAF2 recruitment, and apoptosis pathways in vitro.
  • Eukaryotic expression in HEK293 cells supports proper folding and post-translational modifications.
  • C-terminal 6xHis tag enables facile purification and detection.
  • High affinity for TNF-α, suitable for receptor binding studies.
  • Useful in assays probing apoptosis and TNF receptor signaling.
  • Provided for research use only; not for human or diagnostic use.
Catalog No. 50-001-84831