Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Mo 1mg
SDP
Supplier:  Medchemexpress LLC HYP705161MG
Encompass_Preferred
Recombinant extracellular domain of mouse tumor necrosis factor receptor 2 (TNFRSF1B), expressed in HEK293 cells and supplied with a C-terminal His tag. The protein is purified and quality-controlled for use in biochemical binding studies, functional assays, and reagent development; typical reported purity is ~97.9%.
  • Purified extracellular domain suitable for binding studies and assays.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal His tag enables straightforward purification and detection.
  • Available in multiple sizes to accommodate small-scale and larger experiments.
  • High reported purity supports reliable biochemical and functional results.
  • Provided with QC data such as purity and SDS-PAGE for lot verification.
Catalog No. 50-001-85487