Learn More
Medchemexpress LLC TNF RII/TNFRSF1B Mo 500ug
SDP
Supplier:  Medchemexpress LLC HYP70516500UG
Encompass_Preferred
Recombinant mouse TNF RII (TNFRSF1B) protein produced in HEK293 cells with a C-terminal 6xHis tag. The soluble receptor retains mammalian post-translational modifications and is intended for receptor-ligand binding studies, assay development, and immunology research requiring biologically relevant glycosylation.
  • Mammalian expression preserves native glycosylation and folding.
  • C-terminal 6xHis tag enables straightforward purification and detection.
  • Well suited for receptor-ligand binding assays and ELISA development.
  • Provides a biologically relevant reagent for cell signaling and immunology studies.
Catalog No. 50-001-84324