Learn More
Sino Biological TNF-alpha / TNFA / TNFSF2 Antibody (HRP), Mouse MAb 100ug
Supplier:  Sino Biological 10602MM08H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNF-alpha / TNFA / TNFSF2 (rh TNF-alpha / TNFA / TNFSF2; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-8064