Learn More
Medchemexpress LLC TNIK-IN-8 | 3068941-89-6 | 98.8% | 385.43 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16231410MM/1ML
Encompass_Preferred
TNIK-IN-8 (Compound 35b) is a potent, orally active TNKI inhibitor with an IC50 value of 6 nM. It exhibits antitumor activity.
  • Potent, orally active TNKI inhibitor
  • IC50 value of 6 nM
  • Exhibits antitumor activity
  • Inhibits cell growth in human DLD-1 cells with an IC50 of 21.47 μM
  • Inhibits cell growth in human HCT-116 cells with an IC50 of 2.11 μM
  • Inhibits cell growth in human HT-29 cells with an IC50 of 13.6 μM
Catalog No. 50-003-92829