Learn More
ILEX LIFE SCIENCES LLC PAN Biotech TNM FH Insect Cells Medium with L Glutamine with NaHCO3 500 ML 1/EA
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0480500
Encompass_Preferred
PAN-Biotech TNM-FH Insect Medium w L-Glutamine w Lactalbumin Hydrolysate w Yeast extract w 0 35 g/L NaHC03 500 ml DESCRIPTION Hink s TNM-FH Insect Medium is a modification of Grace s Insect Medium it contains lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate (yeast extract) and was originally developed for the optimal propagation of Tricoplusia ni (cabbage looper) cells The medium can be used to support the growth of a variety of lepidopteran cells BICARBONATE With 0 35 g/L sodium bicarbonate GLUCOSE With glucose GLUTAMINE With L-Glutamine HEPES Without HEPES LIQUID/POWDER Liquid PHENOL RED Without phenol red SODIUM PYRUVATE Without sodium pyruvate STERILE Yes STORAGE TEMP 2 C - 8 C MANUFACTURER PAN-Biotech GmbH COUNTRY OF ORIGIN Germany USAGE This product is intended for Laboratory Research Use Only Not for use in diagnostic or therapeutic procedures
Catalog No. 50-005-80182