Learn More
Bioworld TNM-FH Insect Medium, 500 mL
Supplier:  Bioworld 306110971
Encompass_Preferred
TNM-FH insect medium, also called supplemented Grace’s medium, is ideal for general cell growth and maintenance when supplemented with 2-10% fetal bovine serum. It supports a number of insect cell lines from a variety of lepidopteran species. This medium contains the lactalbumin hydrolysate and yeastolate absent from our Grace’s insect medium. We manufacture all our insect media only with stabilized L-glutamine to ensure the longest possible shelf life.SpecificationsSterile-filteredShips on ice.
Catalog No. 50-254-214
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.