Learn More
TARGETMOL CHEMICALS INC TNO155 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T131765MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TNO155 is a protein tyrosine phosphatase (PTP) non-receptor type 11 (SHP2; src homology region 2 domain phosphatase; PTPN11) inhibitor(IC50 0.011 uM)with potential antineoplastic activity. purity: 99%
Catalog No. 50-258-1373